Would you like to log in again?
BD BBL™ Mueller Hinton II Broth, BD Biosciences
Supplier: BD Biosciences
BBL™
212322 298268 297963 212322 298268
90005-060EA 84.94 USD InStock
90005-060 CA90000-602 CA-90005-112 90000-602 90005-112
BD BBL™ Mueller Hinton II Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
BD BBL™ Mueller Hinton II Broth (Cation-Adjusted) is used for antimicrobial susceptibility testing of microorganisms by broth dilution methods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty