Would you like to log in again?
BBL™ Dermatophyte Test Medium Base, BD Biosciences
Supplier: BD Biosciences
212330 212330
90000-606EA 539.07 USD InStock
CA90000-606 90000-606
BBL™ Dermatophyte Test Medium Base, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Dermatophyte Test Medium Base is a specialized agar used as a highly selective medium for the isolation of dermatophytes.
  • Highly selective medium
  • Helps in the isolation and early recognition
  • For members of the Microsporum species and Trichophyton species
Nondermatophytes can be recognized by the absence of color change. Rapidly growing species may effect a complete color change within three days, while slow growers will change color in a longer time.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty