Would you like to log in again?
BBL™ Middlebrook OADC Enrichment, BD Life Sciences
Supplier: BD Life Sciences
BBL™
212240 212351 211886 212240 211886 212351
90000-418PK 435.05 USD InStock
CA90000-564 CA90000-614 90000-418 90000-614 CA90000-418 90000-564
BBL™ Middlebrook OADC Enrichment, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Middlebrook OADC Enrichment is a supplement recommended for isolation and cultivation of Mycobacteria.
  • Enhances recovery on primary isolation
  • Dextrose is an energy source
  • Catalase destroys toxic peroxides
  • Promotes mycobacterial growth
Supplementation of the agar base is required in order to obtain mycobacterial growth. In the enriched medium, sodium chloride maintains osmotic equilibrium; oleic acid, as well as other long chain fatty acids, can be utilized by tubercle bacilli and plays an important role in the metabolism of mycobacteria; the primary effect of albumin is that of protection of the tubercle bacilli against toxic agents and, therefore, it enhances their recovery on primary isolation; dextrose is an energy source; and catalase destroys toxic peroxides that may be present in the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty