Would you like to log in again?
BBL™ Gram Basic Fuchsin, BD
Supplier: BD Biosciences
212545 212545
90000-716EA 1024.53 USD InStock
CA90000-716 90000-716
BBL™ Gram Basic Fuchsin, BD
Media for Microbiology Dehydrated Media for Microbiology
Gram stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.
  • Differential Gram method
  • Stains more intensely than safranin
Basic fuchsin stains many Gram-negative bacteria more intensely than safranin, making them easier to see.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty