Would you like to log in again?
Difco™ Oxgall, BD Diagnostics
Supplier: BD Diagnostics
212820 212820
90000-728EA 954.12 USD InStock
CA90000-728 90000-728
Difco™ Oxgall, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated fresh bile used in the selection of bile-tolerant bacteria.
  • Inhibits the growth of gram-positive microorganisms
  • Provides a regular supply of bile
  • Used to aid the determination of enteric pathogens
Oxgall is manufactured from large quantities of fresh bile by rapid evaporation of the water content. Bile is composed of fatty acids, bile acids, inorganic salts, sulfates, bile pigments, cholesterol, mucin, lecithin, glycuronicacids, porphyrins, and urea.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty