Would you like to log in again?
Difco™ Bile Salts No. 3, BD Biosciences
Supplier: BD Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
213010 213020 213020
90000-748EA 1044.97 USD InStock
CA90000-746 90000-748 CA90000-748
Difco™ Bile Salts No. 3, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Bile Salts No
  • Selective agents
  • Bile extract
  • Effective at less than 1/3 concentration of bile salts
Bile salts no. 3 contains bile extract standardized to provide inhibitory properties for selective media. It is a modified fraction of bile acid salts, providing a refined bile salt. Bile salts no. 3 is effective at less than one-third concentration of bile salts.
3 are used as selective agents for the isolation of gram-negative microorganisms, inhibiting gram-positive cocci.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty