Would you like to log in again?
Difco™ Modified mTEC Agar, BD
Supplier: BD Biosciences
214880 214880 214884
90000-788EA 2707.48 USD InStock
90000-788 CA90000-796 CA90000-788
Difco™ Modified mTEC Agar, BD
Media for Microbiology Additives and Supplements for Microbiology
Modified mTEC Agar is a selective culture medium used for the chromogenic detection and enumeration of thermotolerant Escherichia coli in water by the membrane filtration technique.
  • Peptone is a source of nitrogen, amino acids, and carbon
  • Agar is the solidifying agent
Sodium lauryl sulfate and sodium desoxycholate are selective against Gram-positive bacteria. Red to magenta colonies may be presumptively identified as
E. coli.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty