Would you like to log in again?
Difco™ MI Agar, BD
Supplier: BD Diagnostics
Difco™
214883 214882 214883
90000-794EA 2457.15 USD InStock
CA90000-794 90000-794 CA90000-792
Difco™ MI Agar, BD
Media for Microbiology Prepared Media for Microbiology
MI Agar is a chromogenic/fluorogenic medium used to detect and enumerate Escherichia coli and total coliforms in drinking water by the membrane filtration technique.
  • Peptone provides nitrogen, carbon, and amino acids
  • Yeast extract provides trace elements, vitamins, and amino acids
  • Lactose is a carbon source and fermentable carbohydrate
  • Sodium chloride maintains osmotic equilibrium
MI Agar is approved for use by certified drinking water laboratories for microbial analysis of potable water. Other uses include recreational, surface or marine water, bottled water, groundwater, well water, treatment plant effluents, water from drinking water distribution line, drinking water source water and possibly foods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty