Would you like to log in again?
Phosphate Buffer, pH 7.2, BD Diagnostics
Supplier: BD Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
214973 257385
90000-834PK 422.46 USD InStock
90000-834 95053-332
Phosphate Buffer, pH 7.2, BD Diagnostics
Buffers Cell Culture Buffers
Phosphate Buffer, pH 7.2 is used for the preparation of dilution blanks for use in the examination of waters, dairy products, foods and other materials.
Phosphate buffer is used in the preparation of dilution blanks for use in microbiological testing rather than unbuffered water in order to standardize this potential variable due to the wide variation in the pH of purified water from multiple sources. Sodium carbonate is a pH regulator.
Certifications: Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance
specifications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty