Would you like to log in again?
BBL™ MI Agar, BD
Supplier: BD Life Sciences
214985 214986
90000-838PK 1318.41 USD InStock
CA90000-840 90000-838
BBL™ MI Agar, BD
Media for Microbiology Prepared Media for Microbiology
MI Agar is a chromogenic/fluorogenic medium used to detect and enumerate Escherichia coli and total coliforms in drinking water by the membrane filtration technique.
  • Peptone is a source of nitrogen, carbon, and amino acids
  • Yeast extract provides trace elements, vitamins, and amino acids
  • Lactose is a fermentable carbohydrate and carbon source
  • Sodium chloride maintains osmotic equillibrium
  • Monopotassium and dipotassium phosphates offer buffering capabilities
  • Sodium lauryl sulfate and sodium desoxycholate are selective against gram-positive bacteria
MI Agar is approved for use by certified drinking water laboratories for microbial analysis of potable water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty