Would you like to log in again?
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Supplier: BD Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Difco™
216010 215510 215510 215530 215920 216010 215530 215520 215520
90000-920EA 200.38 USD InStock
90000-920 CA90000-920 CA90000-904 90000-906 90000-904 CA90000-908 CA90000-918 CA90000-906 90000-908
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Media formulations in dehydrated form is for the convenience of the laboratorian and bioprocessing specialist.
  • Consistent, high-quality performance
  • Conveniently dehydrated
The individual ingredients and the final dehydrated product are carefully quality-controlled to assure consistent, high quality performance and obviate the need for media to be prepared in the laboratory from raw materials. Dehydrated culture media may be offered as 100 g and 500 g bottles, 2 kg and 10 kg containers.
Glucose, D (+), anhydrous, for incorporation into microbiological culture media as a source of energy for bacteria and for the determination of fermentation reactions. It is usually employed at a concentration of 0.5 to 1.0% in fermentation media.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty