Would you like to log in again?
Difco™ Maltose (+), Monohydrate, BD Diagnostics
Supplier: BD Diagnostics
Difco™
216830 216830
90000-932EA 218.4 USD InStock
CA90000-932 90000-932
Difco™ Maltose (+), Monohydrate, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Maltose is a component of starch and glycogen, and is used as a nutrient in culture media.
  • Can be used as a sweetener
  • Mammalian and insect cell culture tested
Maltose can be further hydrolyzed to glucose by the action of maltase, an enzyme commonly found in yeast and many other sources.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty