Would you like to log in again?
Difco™ Soluble Starch, BD Life Sciences
Supplier: BD Life Sciences
217820 217820
90000-952EA 153.79 USD InStock
90000-952 CA90000-952
Difco™ Soluble Starch, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Provides starch for hydrolysis, detoxification of metabolic byproducts, and as a carbon source.
  • Can be purposed to a variety of applications
Soluble starch features a high molecular weight and is a water-soluble dextrin that provides a source of starch for microorganisms to convert into energy. It is also useful in iodimetry, as it gives an easily visible purple-black end point in the presence of free iodine.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty