Would you like to log in again?
Difco™ MIL Medium, BD
Supplier: BD Diagnostics
Difco™
218041 218041
90000-962EA 295.87 USD InStock
CA90000-962 90000-962
Difco™ MIL Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
Motility indole lysine medium is used for differentiating Enterobacteriaceae based on motility, lysine decarboxylation, lysine deamination and indole production.
  • Dehydrated appearance: light beige, free-flowing, homogeneous
  • Prepared appearance: reddish purple, clear, semisolid
  • pH: 6.6 ±0.2 (at 25 °C)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty