Would you like to log in again?
Reinforced Clostridial Medium, BD Diagnostics
Supplier: BD Diagnostics
218081 218081
90000-968EA 237.61 USD InStock
90000-968 CA90000-968
Reinforced Clostridial Medium, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Reinforced Clostridial Medium is used for the cultivation and enumeration of anaerobes, particularly Clostridia, from foods and clinical specimens.
  • Peptone and beef extract are sources of carbon, nitrogen, vitamins, and minerals
  • Sodium acetate acts as a buffer
  • Small amount of agar makes the medium semisolid
  • Dehydrated appearance: Light tan, free-flowing
Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) performance specifications, where applicable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty