Would you like to log in again?
Giolitti-Cantoni Broth Base Dehydrated Culture Media, BD Diagnostics
Supplier: BD Diagnostics
218091 218091
90000-970EA 264.84 USD InStock
CA90000-970 90000-970
Giolitti-Cantoni Broth Base Dehydrated Culture Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Giolitti-Cantoni broth base is used for enriching Staphylococcus aureus from foods during isolation procedures.
It contains peptone and beef extract as sources of carbon, nitrogen, vitamins and minerals. Yeast extract supplies B-complex vitamins which stimulate bacterial growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty