Would you like to log in again?
Difco™ Buffered Peptone Water, Pre-enrichment, BD Life Sciences
Supplier: BD Life Sciences
Difco™
218104 218103 218105
90000-974EA 4475.99 USD InStock
90000-974 90000-972 90000-976
Difco™ Buffered Peptone Water, Pre-enrichment, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Difco™ Buffered Peptone Water, Pre-enrichment is used for preenriching injured Salmonella species from food specimens to increase recovery.
  • Nonselective medium
  • Maintains a high pH over the preenrichment period
These preenrichment media contain peptone as a source of carbon, nitrogen, vitamins and minerals. Sodium chloride maintains the osmotic balance. Phosphates buffer the media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty