Would you like to log in again?
R2A Agar, BD
Supplier: BD Diagnostics
218262 218261 218263 218263
90000-994EA 315.43 USD InStock
CA90000-994 90000-994 CA90000-992 90000-990
R2A Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
R2A Agar is used for enumerating heterotrophic organisms in treated potable water.
  • Dehydrated appearance: Light beige, flree-flowing
  • Prepared appearance: Light amber, slightly opalescent
  • Improves recovery of chlorine-tolerant bacteria
A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty