Would you like to log in again?
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Supplier: BD Diagnostics
218581 218581
90001-010EA 217.39 USD InStock
90001-010 CA90001-010
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Rappaport-Vassiliadis R10 Broth is a liquid medium for selectively enriching Salmonella from meat and dairy products, feces, and polluted water.
  • Tryptone provides carbon and nitrogen for general growth
  • Malachite green is inhibitory to organisms other than Salmonella
  • Low pH helps select for Salmonella species
Selective media recommended by the European Chocolate Manufacturer's Association and the AOAC for rapid detection of motile Salmonella.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty