Would you like to log in again?
Difco™ Brilliant Green Agar, Modified, BD Life Sciences
Supplier: BD Life Sciences
Difco™
218801 218801
90001-022EA 241.25 USD InStock
90001-022 CA90001-022
Difco™ Brilliant Green Agar, Modified, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Brilliant green agar is a selective medium for the isolation of Salmonella from water, sewage and foodstuffs.
  • Dehydrated culture media
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty