Would you like to log in again?
Difco™ MacConkey Broth, BD
Supplier: BD Diagnostics
Difco™
220100 220100
90001-042EA 197.43 USD InStock
90001-042 CA90001-042
Difco™ MacConkey Broth, BD
Media for Microbiology Prepared Media for Microbiology
MacConkey Broth is used for the detection of coliform organisms in milk and water.
  • Peptone provides amino acids and other growth factors
  • Lactose is a carbon energy source
  • Oxgall inhibits the growth of gram-positive organisms
  • Bromcresol purple is the indicator
MacConkey Broth is a modification of the original bile salt broth recommended by MacConkey that contained 0.5% sodium taurocholate and litmus as an indicator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty