Would you like to log in again?
BD BBL™ CultureSwab® EZ Collection and Transport System, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™ CultureSwab®
220144 220145 220144 220145
90001-090PK 198.31 USD InStock
90001-090 90001-092 CA90001-090 CA90001-092
BD BBL™ CultureSwab® EZ Collection and Transport System, BD Diagnostics
Swabs Transport Swabs
Medium-free systems prevent specimen dilution and contamination that can occur with devices containing transport media
Systems feature swabs with a special polyurethane open-celled structure that releases organisms from their environment and protects them during transport. Swabs have a 13.3cm (51/4") long shaft to reach tough sampling sites. They fit into round-bottom transport tubes that stand upright in traditional racks for easy transportation. The caps for the transport tubes are color-coded for easy identification and the double action seal prevents dried out or leaky samples for increased safety.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty