Would you like to log in again?
Difco™ Listeria Enrichment Broth, Modified, BD Life Sciences
Supplier: BD Diagnostics
Difco™
220530 245152 220530 245152
90001-124EA 194.45 USD InStock
90001-124 CA90001-124 90003-376 CA90003-376
Difco™ Listeria Enrichment Broth, Modified, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Listeria Enrichment Broth, Modified is used for selectively enriching Listeria from raw and pasteurized milk according to the International Dairy Federation (IDF).
  • Peptones and yeast extract provide nitrogen, vitamins, and minerals
  • Dextrose is a carbohydrate source
  • Sodium chloride maintains the osmotic balance of the medium
  • Phosphates provide buffering capacity
Listeria Enrichment Broth, Modified is a modification of Listeria Enrichment Broth in which the concentration of one of the selective agents, acriflavine, has been reduced from 15 mg to 10 mg per liter. This modification reflects the lower concentration specified by the IDF for isolation of L. monocytogenes from milk and milk products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty