Would you like to log in again?
BBL™ Kligler Iron Agar (KIA), Slants, BD
Supplier: BD Life Sciences
BBL™
220897 220897
90001-148CS 351.63 USD InStock
90001-148 CA-90001-148
BBL™ Kligler Iron Agar (KIA), Slants, BD
Media for Microbiology Prepared Media for Microbiology
Kligler Iron Agar aids in the differentiation of gram-negative enteric bacilli on the basis of their ability to ferment dextrose and lactose and produce sulfides.
  • Casein and animal tissue provide essential nutrients
  • Hydrogen sulfide production is detected by ferric ammonium citrate and sodium thiosulfate
  • Black color formation is evidence of hydrogen sulfide production
  • Gas production is detected as individual bubbles or displacement of agar
Kligler Iron Agar is used for the differentiation of members of the Enterobacteriaceae on the basis of their ability to ferment dextrose and lactose and to produce sulfides.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty