Would you like to log in again?
BBL™ Lysine Iron Agar (LIA), Slants, BD
Supplier: BD Life Sciences
BBL™
220953
90001-160CS 393.9 USD InStock
90001-160
BBL™ Lysine Iron Agar (LIA), Slants, BD
Media for Microbiology Prepared Media for Microbiology
Lysine Iron Agar is a differential medium for use in the identification of enteric bacilli.
  • Dextrose is a source of fermentable carbohydrate
  • Bromcresol purple is a pH indicator
  • Lysine is the substrate for detecting enzymes
  • Lysine decarboxylase production causes an alkaline or neutral reaction
Lysine Iron Agar is used for the differentiation of enteric organisms based on their ability to decarboxylate or deaminate lysine and to form hydrogen sulfide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty