Would you like to log in again?
BD BBL™ SIM Prepared Tube Media, BD Diagnostics
Supplier: BD Diagnostics
BBL™
221010 221010
90001-180PK 44.36 USD InStock
90001-180 CA90001-180
BD BBL™ SIM Prepared Tube Media, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The SIM prepared tube media is used to differentiate enteric bacilli on the basis of sulfide production, indole formation and motility.
Tube size K.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty