Would you like to log in again?
BBL™ Sabouraud Dextrose Agar (Emmons), Slants, BD
Supplier: BD Life Sciences
221827 221825 221013 221012 221012 221827 221825 221013
90006-488PK 534.74 USD InStock
90006-488 90001-544 CA-90001-186 CA-90001-544 90001-184 90001-186 CA90001-184 CA90006-488
BBL™ Sabouraud Dextrose Agar (Emmons), Slants, BD
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose Agar, Emmons is a nonselective medium for the cultivation and maintenance of fungi.
  • Peptones provide a source of nitrogenous growth factors
  • Dextrose provides an energy source
  • Agar is the solidifying agent
Sabouraud Dextrose Agar, Emmons is a peptone medium supplemented with dextrose to support the growth of fungi.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty