Would you like to log in again?
BBL™ TSI Agar Slants (Triple Sugar Iron Agar), BD
Supplier: BD Life Sciences
221039 221039 221038 221038
90001-206PK 48.34 USD InStock
CA-90001-208 90001-206 CA90001-206 90001-208
BBL™ TSI Agar Slants (Triple Sugar Iron Agar), BD
Media for Microbiology Prepared Media for Microbiology
Triple Sugar Iron Agar (TSI Agar) is used for the differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and the production of hydrogen sulfide.
  • Contains three sugars: dextrose, lactose, and sucrose
  • Phenol red allows indication of carbohydrate fermentation
  • Ferrous ammonium sulfate detects hydrogen sulfide production
To enhance the alkaline condition of the slant, free exchange of air must be permitted by closing the tube cap loosely. If the tube is tightly closed, an acid reaction (caused solely by dextrose fermentation) will also involve the slant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty