Would you like to log in again?
BBL™ Urea Agar, BD Life Sciences
Supplier: BD Life Sciences
221096 221097 221096 221097
90001-228PK 69.18 USD InStock
CA-90001-230 90001-228 CA90001-228 90001-230
BBL™ Urea Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Urea Agar is a differential medium for members of the Enterobacteriaceae on the basis of their ability to produce urease.
  • Peptone and dextrose provide nutrients
  • Reduced buffer content helps promote rapid growth
  • Utilized urea produces ammonia
When organisms utilize urea, ammonia is formed during incubation, which makes the reaction of these media alkaline, producing a pink-red color. Consequently, urease production may be detected by the change in the phenol red indicator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty