Would you like to log in again?
BBL™ Enterococcosel™ Broth, BD Biosciences
Supplier: BD Biosciences
221383 212207 212207
90000-530EA 590.13 USD InStock
CA90000-530 90000-530 90001-342
BBL™ Enterococcosel™ Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Enterococcosel™ Broth is used for the presumptive identification of enterococci.
  • Selective medium
  • Effectively differentiates enterococci
The combination of esculin and a rather low concentration of bile in the presence of azide permits the selection of enterococci and differentiation by esculin hydrolysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty