Would you like to log in again?
BBL™ Bile Esculin Agar Slants, BD
Supplier: BD
BBL™
221410 221409 221409 221410
90001-362PK 51.53 USD InStock
CA-90001-364 90001-362 90001-364 CA90001-362
BBL™ Bile Esculin Agar Slants, BD
Media for Microbiology Prepared Media for Microbiology
Bile Esculin Agar is used to differentiate enterococci and the Streptococcus bovis group from other streptococci.
  • Presumptive identification of streptococci
  • Differentiator medium
Bile Esculin Agar is a medium for the presumptive identification of Enterococcus species and the Streptococcus bovis group of streptococci.
Rochaix noted the value of esculin hydrolysis in the identification of enterococci. Meyer and Schonfeld incorporated bile into the esculin medium and showed that 61 of 62 enterococci were able to grow and split esculin, whereas the other streptococci could not. Swan used an esculin medium containing 40% bile salts and reported that a positive reaction on the bile esculin medium correlated with a serological group D precipitin reaction.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty