Would you like to log in again?
BBL™ Lowenstein-Jensen Medium Slants, BD Life Sciences
Supplier: BD Life Sciences
BBL™
220909 220908 221414 220908 220909
90001-368CS 598.5 USD InStock
CA90001-156 90001-368 90001-154 CA90001-154 90001-156
BBL™ Lowenstein-Jensen Medium Slants, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
BBL™ L-J Medium is a selective medium for the isolation of mycobacteria from specimens containing mixed flora.
  • Contains inorganic salts and other sources of nitrogen
  • Glycerol provides source of carbon and energy
  • Egg yolk is a source of lipids
  • Malachite green dye partially inhibits growth of other bacteria
BD BBL™ Mycobactosel™ L-J Medium is Lowenstein-Jensen Medium1-3 plus cycloheximide, lincomycin and nalidixic acid for use with specimens likely to contain many contaminating organisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty