Would you like to log in again?
Difco™ Salmonella O Antigen and Antisera Groups, BD
Supplier: BD Diagnostics
BD Taxo™
222591 222591 211783 221421 221321 211778 221301 211780 211781 211778 211753 221301 221421 221341
90000-356EA 538.04 USD InStock
90000-356 CA90000-326 90001-370 CA90001-370 90001-294 CA90001-294 90001-662 CA90001-662 90001-324 CA90001-302 90000-350 90000-354 CA90000-350 90000-352
Difco™ Salmonella O Antigen and Antisera Groups, BD
Media for Microbiology Dehydrated Media for Microbiology
BD Difco™ QC Antigens Salmonella O and BD Difco QC Antigen Salmonella Vi are used in the quality control testing of BD Difco Salmonella O Antisera and BD Difco Salmonella Vi Antiserum by slide agglutination tests.
  • May be used to demonstrate a negative control reaction
  • Chemically stable
  • Inactivated suspension
These antigens are chemically stabilized and inactivated suspensions of known strains of the genus Salmonella designed for use as homologous controls for testing the efficacy of the Salmonella typing antisera employed in routine laboratory procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty