Would you like to log in again?
BBL™ Cooked Meat Medium, BD
Supplier: BD Biosciences
BBL™
221508 221508
90001-374PK 689.11 USD InStock
CA-90001-374 90001-374
BBL™ Cooked Meat Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
BBL™ cooked meat medium is used for the cultivation of anaerobes in particular clostridia pathogens.
  • Supports the growth of most of sporulated, non-spore-bound anaerobes
  • Muscle protein present in the granules of cardiac tissue is a source of amino acids and others nutrients
  • Growth is indicated by turbidity and presence of gas bubbles
Robertson developed a ground with cooked meat to use in the cultivation of some anaerobes isolated from wounds. The current formulation of the ground with cooked meat is a modification of the Robertson's original formula.
Can be used for the cultivation and conservation of clostridia and for the determination of the proteolytic activity of anaerobes.
This medium provides a favorable environment for the growth of anaerobes. The sulfhydryl groups, which exert the reducing effect, are more available in denatured protein; therefore, the meat particles are cooked for use in the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty