Would you like to log in again?
BBL™ Phenol Red Broth, BD Life Sciences
Supplier: BD Life Sciences
BBL™
221673 221673 221691 221687 221677 221687 221699 221699 221677 221689
90001-454PK 96.14 USD InStock
90001-454 CA90001-454 90001-468 90001-458 CA90001-468 CA90001-458 90001-480 CA90001-480 90001-470 90001-472
BBL™ Phenol Red Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Phenol red broth with carbohydrates are used for the determination of fermentation reactions in the differentiation of microorganisms.
  • Complete medium
  • Detect acid production
  • Used as negative control for fermentation
Most of the end products of carbohydrate fermentation are organic acids which, in the presence of phenol red, produce a color change in the medium from red to yellow.
If gas is produced during the fermentation reaction, it is collected in the inverted Durham tube.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty