Would you like to log in again?
BBL™ OF Medium, BD Life Sciences
Supplier: BD Life Sciences
BBL™
221518 221705 221517 221705 221703 221703 221714 221517 221328 221328 221714 297366
90004-856PK 54.65 USD InStock
CA90001-486 90004-856 90001-380 90001-308 90001-382 90001-492 CA90001-380 CA90001-308 90001-484 CA90001-492 CA90001-484 90001-486
BBL™ OF Medium, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
OF Basal Medium, when supplemented with an appropriate carbohydrate, is used to determine the oxidative and fermentative metabolic activities of gram-negative bacilli.
  • Carbohydrate provides a source of energy
  • Dipotassium phosphate adds buffering capacity to the medium
  • Agar permits determination of motility
Changed in the covered agar are considered to be due to true fermentation, while changes in the open tubes are due to oxidative utilization of the carbohydrate present. If the carbohydrate is not utilized by either method, there is no acid production in either tube.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty