Would you like to log in again?
BBL™ Middlebrook 7H9 Broth with Glycerol, BD Life Sciences
Supplier: BD Life Sciences
BBL™
221832 221832
90001-552PK 126.88 USD InStock
CA90001-552 90001-552
BBL™ Middlebrook 7H9 Broth with Glycerol, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Middlebrook 7H9 Broth dehydrated base supplemented with glycerol supports the growth of mycobacteria, including M
  • Inorganic salts provides essential growth substances
  • Sodium citrate holds certain inorganic cations in solution
  • Glycerol provides carbon and energy
  • Malachite green dye partially inhibits bacterial contaminants
Middlebrook 7H9 Broth with appropriate supplements is primarily used for growth of pure cultures of mycobacteria for use in laboratory studies.
tuberculosis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty