Would you like to log in again?
BD BBL™ Herrold’s Egg Yolk Agar, BD Diagnostics
Supplier: BD Diagnostics
BBL™
222241 222232 222233 222240
90001-620PK 102.36 USD InStock
90001-620 90001-622 90001-624 90001-626
BD BBL™ Herrold’s Egg Yolk Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Herrold’s egg yolk agar contains amphotericin B, nalidixic acid and vancomycin
  • Selective isolation of Mycobacterium paratuberculosis
  • Helps Mycobacterial growth
Agar without mycobactin helps in the growth of Mycobacteria other than Mycobacterium paratuberculosis.
It is used for the selective isolation and differentiation of Mycobacterium paratuberculosis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty