Would you like to log in again?
Difco™ Malt Agar, BD Diagnostics
Supplier: BD Diagnostics
Difco™
224200 224100 224200 224100
90001-762EA 7480.53 USD InStock
CA90001-772 90001-762 CA90001-762 90001-772
Difco™ Malt Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malt Agar is used for isolating and cultivating yeasts and molds from food and for cultivating yeast and mold stock cultures.
  • Malt extract provides carbon, protein, and essential nutrients
  • Acidic pH level
  • Agar is the solidifying agent
The acidic pH of Malt Agar allows for optimal growth of molds and yeasts while restricting bacterial growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty