Would you like to log in again?
Dehydrated Antibiotic Medium 3, BD Life Sciences
Supplier: BD Life Sciences
210932 224320 224320 210932
90000-024EA 214.05 USD InStock
CA90000-024 90000-024 90001-780 CA90001-780
Dehydrated Antibiotic Medium 3, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated antibiotic medium number 3 is used for the turbidimetric assay of antibiotics.
  • Dehydrated
  • Used for turbidimetric assay of antibiotics
Antibiotic Medium 3 can also be used for high-quality fungal testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty