Would you like to log in again?
Difco™ Sodium Desoxycholate, BD Life Sciences
Supplier: BD Life Sciences
224820 224820
90001-806EA 474.71 USD InStock
90001-806 CA90001-806
Difco™ Sodium Desoxycholate, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Sodium salt of desoxycholic acid used for the pneumococcus bile solubility test and as a media component.
  • Popular anionic, bile-acid detergent
  • Can be removed from solution by dialysis
  • High-purity compound with low UV absorptivity
Sodium desoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty