Would you like to log in again?
Difco™ Triple Sugar Iron Agar, BD Biosciences
Supplier: BD Biosciences
226540 226540
90001-896EA 170.95 USD InStock
90001-896 CA90001-896
Difco™ Triple Sugar Iron Agar, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Triple Sugar Iron Agar (TSI Agar) is used for the differentiation of Gram-negative enteric bacilli based on carbohydrate fermentation and the production of hydrogen sulfide.
  • Contains three sugar: dextrose, lactose, and sucrose
  • Phenol red for detecting carbohydrate fermentation
  • Ferrous ammonium sulfate detects hydrogen sulfide production
  • Dehydrated appearance: Pink, free-flowing
The addition of sucrose to the double sugar formulation of Kligler Iron Agar increased the sensitivity of the medium by facilitating the detection of sucrose-fermenting bacilli, as well as lactose and/or dextrose fermenters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty