Would you like to log in again?
Difco™ Cooked Meat Medium, BD
Supplier: BD Life Sciences
226730 226730
90001-914EA 1148.91 USD InStock
90001-914 CA90001-914
Difco™ Cooked Meat Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
Cooked Meat Medium and the enriched medium are used for the cultivation of anaerobes, especially pathogenic clostridia.
  • Muscle protein provides amino acids and other nutrients
  • Muscle provides reducing substances to permit growth of strict anaerobes
  • Growth is indicated by turbidity and presence of gas bubbles
Cooked Meat Medium provides a favorable environment for the growth of anaerobes. The sulfhydryl groups, which exert the reducing effect, are more available in denatured protein; therefore, the meat particles are cooked for use in the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty