Would you like to log in again?
Difco™ Nitrate Broth, BD Life Sciences
Supplier: BD Life Sciences
Difco™
226810 226810
90001-916EA 521.03 USD InStock
90001-916 CA90001-916
Difco™ Nitrate Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Nitrate Broth is recommended as an aid in the identification of aerobic and facultative anaerobic gram-negative microorganisms by means of the nitrate reduction test.
  • Basal medium containing potassium nitrate
  • Addition of zinc dust is used to detect unreduced nitrate
  • Organisms derive oxygen from nitrate through anaerobic respiration
The formation of nitrogen gas, an end product typical of certain organisms, is evidenced by displacement of the medium from the Durham tube by the gas produced.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty