Would you like to log in again?
Difco™ Tryptic Soy Blood Agar Base, BD Biosciences
Supplier: BD Biosciences
227300 227200 228200 227300 227200
90001-922EA 2619.33 USD InStock
90001-922 CA90001-922 90001-926 CA90001-926 90001-970
Difco™ Tryptic Soy Blood Agar Base, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tryptic Soy Blood Agar Base No
  • Clear hemolytic reactions with group A streptococci
  • Tryptic Soy Blood Agar base EH provides dramatic, improved hemolysis
  • Specified in standard methods for food testing
Tryptic Soy Blood Agar Base No. 2 and Tryptic Soy Blood Agar Blase EH are similar in composition to TSA II. These formulations utilize the peptones Tryptone H and Tryptone H Plus to enhance hemolysin production while minimizing antagonism or loss in activity of streptococcal hemolysins.
2 and Tryptic Soy Blood Agar Base EH are used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty