Would you like to log in again?
Difco™ Desoxycholate Agar, BD Life Sciences
Supplier: BD Life Sciences
227310 227310
90001-928EA 198.51 USD InStock
90001-928 CA90001-928
Difco™ Desoxycholate Agar, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Desoxycholate Agar is a slightly selective and differential plating medium used for isolating and differentiating gram-negative enteric bacilli.
  • Peptone provides nitrogen and carbon
  • Lactose is the fermentable carbohydrate
  • Sodium chloride and dipotassium phosphate maintain osmotic equilibrium
  • Sodium desoxycholate inhibits gram-positive bacteria
  • Ferric citrate and sodium citrate also inhibit growth
  • Neutral red is a pH indicator
  • Agar is the solidifying agent
Differentiation of enteric bacilli is based on fermentation of lactose. Bacteria that ferment lactose produce acid and, in the presence of neutral red, form red colonies. Bacteria that do not ferment lactose form colorless colonies. The majority of normal intestinal bacteria ferment lactose, while Salmonella and Shigella species do not ferment lactose.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty