Would you like to log in again?
Difco™ Supplement B with Reconstituting Fluid, BD Biosciences
Supplier: BD Biosciences
227610 227610
90001-934PK 465.36 USD InStock
90001-934 CA90001-934
Difco™ Supplement B with Reconstituting Fluid, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Supplement B with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
  • Contains yeast concentrate, glutamine, coenzyme, cocarboxylase, hematin, and growth factors
  • For use in augmenting the preparation of chocolate agar
Supplement B is a yeast concentrate for use in supplementing media for growth of microorganisms with exacting nutritional requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty