Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Diagnostic Tests—Adjuvant Complete to Neisseria meningitidis Antiserum, BD Diagnostics
Supplier: BD Diagnostics
BD Difco™
222501 223101 229701 222501 222321 227921 240661 222371 222361 222291 229101 225851 222521 223021 240658 229701 227911 223041 223101 228811 231131 222281 231141 227901 240661 263810 221591 227891 223091 223011 222521 222301 223143 222531 222281 221591 228911 223143 222311 240658 228801 222301 222531 225851 228811 223142 264010 227891 263910 264010 222311 228801 222371 223021 222361 263910 222321 227911 223091 231141 231131 222291 229101 223001 227921 223142 263810 228911 223031 227901
90002-020EA 395.36 USD InStock
90002-020 90003-746 90003-748 CA90001-390 90001-652 90001-732 90001-656 CA90001-742 90001-736 CA90001-744 CA90001-628 CA90001-948 CA90003-748 CA90002-040 CA90003-746 90001-390 CA90010-142 90001-742 CA90001-950 CA90001-630 CA90001-952 90001-744 CA90001-632 CA90001-956 90001-628 CA90001-636 90001-948 CA90001-718 CA90002-010 90002-040 90003-126 90003-124 90001-882 90001-640 CA90001-640 90001-630 90001-950 90001-632 CA90010-128 90001-952 CA90001-644 CA90002-208 90001-712 CA90001-646 90001-636 90001-956 90001-714 CA90002-206 CA90001-648 CA90002-004 90001-718 CA90002-002 CA90002-020 90002-010 CA90003-124 CA90003-126 90003-756 CA90001-652 90001-644 CA90001-732 90001-722 90001-646 90001-724 90002-208 90001-648 CA90001-656 CA90001-736 90002-206 90002-004 90002-002
Diagnostic Tests—Adjuvant Complete to Neisseria meningitidis Antiserum, BD Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Antigens and antisera used for slide agglutination tests and immunofluorescence. BD Difco Listeria O Antigens Types 1 and 4 (Slide) are suspensions of appropriate serotypes containing 0.3% formaldehyde as a preservative
Store at 2 to 8 °C (35.6 to 46.4 °F).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty