Would you like to log in again?
Difco™ Brilliant Green Agar, BD Life Sciences
Supplier: BD Life Sciences
228530 228530
90002-000EA 247.73 USD InStock
90002-000 CA90002-000
Difco™ Brilliant Green Agar, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S
  • Brilliant green dye inhibits gram-positive bacteria
  • Inhibits most gram-negative bacilli
  • Phenol red serves as a pH indicator
Yellow coloring appears as a result of acid production in the fermentation of lactose and/or sucrose in the medium.
Typhi from feces and other materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty