Would you like to log in again?
Difco™ Mitis Salivarius Agar, BD
Supplier: BD Diagnostics
Difco™
229810 229810 293093
90008-248PK 185.5 USD InStock
90008-248 CA90002-046 90002-046
Difco™ Mitis Salivarius Agar, BD
Media for Microbiology Prepared Media for Microbiology
Mitis Salivarius Agar is used with Tellurite Solution 1% in isolating Streptococcus mitis.
  • Peptones as a source of carbon, nitrogen, and nutrients
  • Dextrose and saccharose provide carbohydrate sources
  • Crystal violet and potassium tellurite inhibit bacterial growth
  • Tryptan blue gives colonies a blue color
  • Agar is the solidifying agent
S. mitis and S. salivarius are known as viridans streptococci, and these organisms play a role in cariogenesis and infective endocarditis and cause an increasing number of bacteremias.
Salivarius, and enterococci, particularly from grossly contaminated specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty